missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Sodium Tartrate, 4-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Specifications
Specifications
| Physical Form | Crystals |
| Quantity | 1 kg |
| Formula Weight | 282.22 |
| Percent Purity | 99.9% |
| Grade | BAKER ANALYZED™ ACS Reagent |
| Packaging | Poly Bottle |
| Specific Gravity | 1L = 1.79kg |
| Chemical Name or Material | Potassium Sodium Tartrate, 4-Hydrate |