missing translation for 'onlineSavingsMsg'
Learn More

Potassium Sodium Tartrate, 4-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

High quality chemicals for laboratory and specialized industrial use

Supplier:  Avantor J.T.Baker 326205

 View more versions of this product

Catalog No. 02003831

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

Potassium Sodium Tartrate
6381-59-5
2.5 kg
potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j
C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+]
282.218
282.22
Poly Bottle
4-Hydrate
Crystals
C4H12KNaO10
VZOPRCCTKLAGPN-UHFFFAOYSA-L
potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate
2724148
BAKER ANALYZED™ ACS Reagent
1L = 1.79kg

Chemical Identifiers

6381-59-5
282.218
potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j
potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate
C4H12KNaO10
VZOPRCCTKLAGPN-UHFFFAOYSA-L
2724148
C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+]
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions