missing translation for 'onlineSavingsMsg'
Learn More

Potassium selenate, 99.5% (metals basis)

Catalog No. p-4407761
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA6811104 2 g
AA6811109 10 g
AA6811118 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA6811104 Supplier Thermo Scientific Chemicals Supplier No. 06811104

CAS: 7790-59-2 | K2O4Se | 221.164 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7790-59-2
Molecular Formula K2O4Se
Molecular Weight (g/mol) 221.164
MDL Number MFCD00054125
InChI Key YAZJAPBTUDGMKO-UHFFFAOYSA-L
Synonym potassium selenate, dipotassium selenate, unii-99798pu7mi, selenic acid, dipotassium salt, 2k.so4e, selenic acid solution, selenic acid, potassium salt 1:2, selenic acid solution, 40 wt. % in water 100g
PubChem CID 62680
IUPAC Name dipotassium;selenate
SMILES [O-][Se](=O)(=O)[O-].[K+].[K+]

Specifications

Color White
Physical Form Powder
Quantity 2 g
Assay Percent Range 99.5% (metals basis)
Solubility Information Soluble in water
Formula Weight 221.16
Odor Odorless
Sensitivity Hygroscopic
Density 3.07 g/cm3 at 20°C
Chemical Name or Material Potassium selenate
Hazard Category H301+H331-H373
Precautionary Statement P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: I
EINECSNumber 232-214-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only