missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium pyrosulfate, pure, Thermo Scientific™
Specifications
Specifications
| Melting Point | 325°C |
| Color | White |
| Physical Form | Adhering Crystalline Powder |
| Quantity | 2.5 kg |
| Packaging | Plastic Bottle |
| Linear Formula | K2S2O7 |
| Merck Index | 13-7749 |
| Solubility Information | May decompose in water |
| Chemical Name or Material | Potassium pyrosulfate |
Safety and Handling
| Hazard Category | Corrosion |
| Hazard Statement | Skull and crossbones |
| EINECSNumber | 232-216-8 |
| Recommended Storage | Normal conditions |