Learn More
Potassium Pyrosulfate, ACS Grade, LabChem™
Supplier: LabChem LC200981
Specifications
| Potassium Pyrosulfate | |
| White | |
| 100 | |
| 500 g | |
| UN3260 | |
| Soluble in water | |
| [O-]S(=O)(=O)OS(=O)(=O)[O-].[K+].[K+] | |
| 254.31 | |
| ACS | |
| Amber Glass |
| 325°C | |
| 7790-62-7 | |
| Powder | |
| K2O7S2 | |
| potassium pyrosulfate, dipotassium disulfate, potassium disulfate, unii-avj6zst7l6, avj6zst7l6, disulfuric acid dipotassium salt, dipotassium sulfonato sulfate, dipotassium disulphate, dipotassium pyrosulfate, kaliumpyrosulfat | |
| KAQHZJVQFBJKCK-UHFFFAOYSA-L | |
| dipotassium;sulfonato sulfate | |
| 62681 | |
| Passes Test | |
| Sulfur compounds |
Chemical Identifiers
| 7790-62-7 | |
| 254.31 | |
| potassium pyrosulfate, dipotassium disulfate, potassium disulfate, unii-avj6zst7l6, avj6zst7l6, disulfuric acid dipotassium salt, dipotassium sulfonato sulfate, dipotassium disulphate, dipotassium pyrosulfate, kaliumpyrosulfat | |
| dipotassium;sulfonato sulfate |
| K2O7S2 | |
| KAQHZJVQFBJKCK-UHFFFAOYSA-L | |
| 62681 | |
| [O-]S(=O)(=O)OS(=O)(=O)[O-].[K+].[K+] |
Safety and Handling
GHS H Statement
May be corrosive to metals.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
GHS P Statement
Keep only in original conatainer.
Do not breathe dust.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Use only outdoors or in a well-ventilated area.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Absorb spillage to prevent material damage.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Store in corrosive resistant container with a resistant inner liner.
Dispose of contents/container to comply with local, state and federal regulations.
Danger
Recommended Storage : Room Temperature