missing translation for 'onlineSavingsMsg'
Learn More

Potassium pyrosulfate, 98%, for analysis

Catalog No. p-4509909
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog No. Quantity Packaging
AC207915000 500 g Plastic bottle
AC207910025 2.5 kg Plastic bottle
2 options

Catalog No. AC207915000

Supplier: Thermo Scientific Chemicals 207915000

Only null left

CAS: 7790-62-7 | K2O7S2 | 254.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis

Chemical Identifiers

CAS 7790-62-7
Molecular Formula K2O7S2
Molecular Weight (g/mol) 254.31
InChI Key KAQHZJVQFBJKCK-UHFFFAOYSA-L
Synonym potassium pyrosulfate, dipotassium disulfate, potassium disulfate, unii-avj6zst7l6, avj6zst7l6, disulfuric acid dipotassium salt, dipotassium sulfonato sulfate, dipotassium disulphate, dipotassium pyrosulfate, kaliumpyrosulfat
PubChem CID 62681
IUPAC Name dipotassium;sulfonato sulfate
SMILES [O-]S(=O)(=O)OS(=O)(=O)[O-].[K+].[K+]

Specifications

Melting Point 325.0°C
Color White
Physical Form Fine Crystalline Powder
Quantity 500 g
Assay Percent Range 98%
Linear Formula K2S2O7
Merck Index 15,7784
Solubility Information Solubility in water: may decompose
Formula Weight 254.31
Percent Purity 98%
Grade Analytical
Packaging Plastic bottle
Specific Gravity 2.51
Density 2.5100g/mL
Chemical Name or Material Potassium pyrosulfate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Corrosive to the respiratory tract.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
EINECSNumber 232-216-8

RUO – Research Use Only