missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Pyrogallate, Orsat, Ricca Chemical
Supplier: Ricca Chemical Company 646032
Specifications
| Potassium Pyrogallate | |
| 51.0,38.96,7.68 | |
| dihydrogen oxide, dihydrogen monoxide | |
| 1 L |
| 7732-18-5,1310-58-3,87-66-1 | |
| 53.0,40.55,8.0 | |
| Amber Poly Bottle |
Chemical Identifiers
| 7732-18-5 | |
| dihydrogen oxide, dihydrogen monoxide |