missing translation for 'onlineSavingsMsg'
Learn More

Potassium Pyroantimonate, Reagent, Reagents

Catalog No. 01337795
Change view
Click to view available options
Quantity: 100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01337795 Quantity: 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01337795 Supplier Labchem Inc Supplier No. C1290000100C2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Potassium Pyroantimonate, Reagent, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 12208-13-8
Molecular Formula H6KO6Sb
Molecular Weight (g/mol) 262.90
InChI Key IDVSLFDKDQVCAP-UHFFFAOYSA-H
Synonym Potassium Hexahydroxyantimonate
IUPAC Name antimony(3+) potassium hexahydroxide
SMILES [OH-].[OH-].[OH-].[OH-].[OH-].[OH-].[K+].[Sb+3]

Specifications

Color White
pH 7.5 to 9.0
Physical Form Solid
Quantity 100 g
Grade Reagent
Packaging Glass Bottle
Specific Gravity 3.34
Chemical Name or Material Potassium Pyroantimonate, Reagent