missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Phthalate ACS AR Crystal, Macron Fine Chemicals™
macron
$4,127.87
Chemical Identifiers
| CAS | 877-24-7 |
|---|---|
| Molecular Formula | C8H5KO4 |
| Molecular Weight (g/mol) | 204.222 |
| InChI Key | IWZKICVEHNUQTL-UHFFFAOYSA-M |
| Synonym | potassium hydrogen phthalate, potassium biphthalate, monopotassium phthalate, potassium acid phthalate, hydrogen potassium phthalate, phthalic acid monopotassium salt, 1,2-benzenedicarboxylic acid, monopotassium salt, phthalic acid, monopotassium salt, phthalic acid potassium salt, potassium phthalate monobasic |
| PubChem CID | 23676735 |
| IUPAC Name | potassium;2-carboxybenzoate |
| SMILES | C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+] |
Chemical Identifiers
| 877-24-7 | |
| 204.222 | |
| potassium hydrogen phthalate, potassium biphthalate, monopotassium phthalate, potassium acid phthalate, hydrogen potassium phthalate, phthalic acid monopotassium salt, 1,2-benzenedicarboxylic acid, monopotassium salt, phthalic acid, monopotassium salt, phthalic acid potassium salt, potassium phthalate monobasic | |
| potassium;2-carboxybenzoate |
| C8H5KO4 | |
| IWZKICVEHNUQTL-UHFFFAOYSA-M | |
| 23676735 | |
| C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+] |
Specifications
| 877-24-7 | |
| potassium hydrogen phthalate, potassium biphthalate, monopotassium phthalate, potassium acid phthalate, hydrogen potassium phthalate, phthalic acid monopotassium salt, 1,2-benzenedicarboxylic acid, monopotassium salt, phthalic acid, monopotassium salt, phthalic acid potassium salt, potassium phthalate monobasic | |
| C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+] | |
| 204.222 | |
| 204.22 | |
| Potassium Phthalate, Primary Standard |
| C8H5KO4 | |
| IWZKICVEHNUQTL-UHFFFAOYSA-M | |
| potassium;2-carboxybenzoate | |
| 23676735 | |
| ACS |