missing translation for 'onlineSavingsMsg'
Learn More

Potassium phosphate, tribasic monohydrate, 96%, extra pure

Catalog No. AC263790250
Change view
Click to view available options
Quantity:
25 g
500 g
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Glass bottle
Plastic Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC263790250 25 g Glass bottle
AC263795000 500 g Glass bottle
AC263790010 1 kg Glass bottle
AC263790050 5 kg Plastic Bottle
AC263790251 25 kg Fibre drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AC263790250 Supplier Thermo Scientific Chemicals Supplier No. 263790250
Only null left

CAS: 27176-10-9 | K3O4P·H2O | 230.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 27176-10-9
Molecular Formula K3O4P·H2O
Molecular Weight (g/mol) 230.28
MDL Number MFCD00150410
InChI Key RMNIZOOYFMNEJJ-UHFFFAOYSA-K
Synonym tri-potassium phosphate monohydrate, potassium phosphate tribasic monohydrate, potassium phosphate hydrate, potassium phosphate monohydrate, unii-4927m96tfv, tert-potassium phosphate monohydrate, potassium phosphate, tribasic, monohydrate, phosphoric acid, tripotassium salt, monohydrate, tripotassium phosphate, monohydrate, acmc-1chx7
PubChem CID 22031701
IUPAC Name tripotassium;phosphate;hydrate
SMILES O.[O-]P(=O)([O-])[O-].[K+].[K+].[K+]

Specifications

Melting Point 1340.0°C
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 96%
Linear Formula K3PO4·H2O
Solubility Information Solubility in water: soluble. Other solubilities: insoluble in alcohol
Formula Weight 230.28
Percent Purity 96%
Grade Extra Pure
Loss on Ignition 13% max. (800°C)
Packaging Glass bottle
Density 2.564
Chemical Name or Material Potassium phosphate, tribasic monohydrate, c.p.
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mis

RUO – Research Use Only