missing translation for 'onlineSavingsMsg'
Learn More

Potassium Phosphate Tribasic, FCC, 97%, Spectrum™ Chemical

Catalog No. 18605683
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
20 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605683 125 g Poly Bottle
18605684 500 g Poly Bottle
18605685 2.5 kg Poly Pail
18605686 12 kg Fiber Drum
18605688 20 kg Fiber Drum
18605687 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18605683 Supplier Spectrum Chemical Mfg Cor Supplier No. P1379125GM
Get Up to $500 in Chemicals with Purchase
View all promotions

P1379, 7778-53-2, K3PO4

Spectrum™ Chemical Potassium Phosphate Tribasic, FCC is used as a food additive for its properties as an emulsifier, foaming agent, and whipping agent. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage, nutritional supplement and personal care applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7778-53-2
Molecular Formula K3O4P
Molecular Weight (g/mol) 212.27
InChI Key LWIHDJKSTIGBAC-UHFFFAOYSA-K
IUPAC Name tripotassium phosphate
SMILES [K+].[K+].[K+].[O-]P([O-])([O-])=O

Specifications

CAS Max % 1
Quantity 125 g
Percent Purity 97%
Grade FCC
Loss on Ignition 0.05
Packaging Poly Bottle