Learn More
Potassium phosphate tribasic, 90+%, pure, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 467330010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| Potassium phosphate tribasic | |
| 1 kg | |
| K3O4P | |
| Solubility aq. soln.: 508 g/L (25°C). Other solubilities: insoluble in alcohol | |
| [K+].[K+].[K+].[O-]P([O-])([O-])=O | |
| 212.27 | |
| Pure |
| 7778-53-2 | |
| 90+% | |
| K3PO4 | |
| LWIHDJKSTIGBAC-UHFFFAOYSA-K | |
| tripotassium phosphate | |
| 212.27 |
Chemical Identifiers
| 7778-53-2 | |
| 212.27 | |
| tripotassium phosphate |
| K3O4P | |
| LWIHDJKSTIGBAC-UHFFFAOYSA-K | |
| [K+].[K+].[K+].[O-]P([O-])([O-])=O |
Safety and Handling
GHS H Statement Causes serious eye damage.
May cause respiratory irritation.
GHS P Statement IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
GHS Signal Word: Danger
EINECSNumber : 231-907-1
RUO â Research Use Only