missing translation for 'onlineSavingsMsg'
Learn More

Potassium phosphate tribasic, 90+%, pure

Catalog No. AC467330250
Change view
Click to view available options
Quantity:
1 kg
5 kg
25 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC467330250 25 kg
AC467330010 1 kg
AC467330050 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC467330250 Supplier Thermo Scientific Chemicals Supplier No. 467330250

CAS: 7778-53-2 | K3O4P | 212.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7778-53-2
Molecular Formula K3O4P
Molecular Weight (g/mol) 212.27
InChI Key LWIHDJKSTIGBAC-UHFFFAOYSA-K
IUPAC Name tripotassium phosphate
SMILES [K+].[K+].[K+].[O-]P([O-])([O-])=O

Specifications

Melting Point 1340.0°C
Quantity 25 kg
Assay Percent Range 90+%
Linear Formula K3PO4
Solubility Information Solubility aq. soln.: 508 g/L (25°C). Other solubilities: insoluble in alcohol
Formula Weight 212.27
Grade Pure
Chemical Name or Material Potassium phosphate tribasic
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Causes serious eye damage.

May cause respiratory irritation.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

Immediately call a POISON CENTER or doctor/physician.

Avoid breathing dust/fume/gas/mist/vapors/spray.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

EINECSNumber 231-907-1

RUO – Research Use Only