accessibility menu, dialog, popup
UserName
Fisher Scientific Logo
EN
chevron-down
Select Language
English
Français
Search
Order Status
Quick Order
Support
Offers
chevron-down
Featured
Spotlight
Offers
Flex Dollars
New Lab Start-up
View All
Sign In
chevron-down
chevron-down
Sign In
Don't have an account?
Create an Account
missing translation for 'onlineSavingsMsg'
Learn More
Home
Products
Gel Electrophoresis Equipment and Supplies
Gel Electrophoresis Reagents and Buffers
Gel Electrophoresis Buffers and Diluents
Potassium Phosphate pH 6.8 DILUT-IT™ Dissolution Media Concentrate, J.T. Baker™
Print
Potassium Phosphate pH 6.8 DILUT-IT™ Dissolution Media Concentrate, J.T. Baker™
Catalog No.
02003694
Change view
Chip view icon
List view icon
Click to view available options
Quantity:
230.8 mL
961.5 mL
1.9 L
Packaging:
Narrow Mouth PE Bottle
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
Quantity
Packaging
02003694
230.8 mL
Poly Bottle
02003695
961.5 mL
Narrow Mouth PE Bottle
02003693
1.9 L
Narrow Mouth PE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No.
02003694
Supplier
Avantor
Supplier No.
D01400
Sign In
or
Register
to check your price and availability.
$9,417.78
/
Case of 12 Each
Add to Cart
Add to Cart
Description
Specifications
Product Suggestions
Videos
Documents
Certificates
Safety and Handling
Safety Services
Promotions
Specifications
Specifications
Quantity
230.8 mL
Packaging
Poly Bottle
Product Suggestions
Documents