missing translation for 'onlineSavingsMsg'
Learn More

Potassium phosphate monohydrate, 95%

Catalog No. AA4158636
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4158636 500 g
AA41586A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4158636 Supplier Thermo Scientific Chemicals Supplier No. 04158636
Only null left

CAS: 27176-10-9 | H2K3O5P | 230.28 g/mol

Potassium phosphate monohydrate acts as a reagent and buffer used in molecular biology, biochemistry, and chromatography. It is utilized to study the effects of freezing and thawing of proteins sensitive to conformational changes. It is also used in the extraction of keratohyalin protein from bovine tissue. Further, it is used as a food additive, emulsifier, foaming agent and fertilizer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 27176-10-9
Molecular Formula H2K3O5P
Molecular Weight (g/mol) 230.28
MDL Number MFCD00150410
InChI Key RMNIZOOYFMNEJJ-UHFFFAOYSA-K
Synonym tri-potassium phosphate monohydrate, potassium phosphate tribasic monohydrate, potassium phosphate hydrate, potassium phosphate monohydrate, unii-4927m96tfv, tert-potassium phosphate monohydrate, potassium phosphate, tribasic, monohydrate, phosphoric acid, tripotassium salt, monohydrate, tripotassium phosphate, monohydrate, acmc-1chx7
PubChem CID 22031701
IUPAC Name tripotassium;phosphate;hydrate
SMILES O.[O-]P(=O)([O-])[O-].[K+].[K+].[K+]

Specifications

Color White
Physical Form Granules
Quantity 500 g
Solubility Information Soluble in water. Insoluble in alcohol.
Formula Weight 230.28 (212.27 Anhydrous)
Odor Odorless
Appearance White granules
Density 2.564
Chemical Name or Material Potassium phosphate monohydrate
Hazard Category H318-H335
Precautionary Statement P261-P271-P280-P304+P340-P305+P351+P338-P310-P312-P501c
EINECSNumber 231-907-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only