missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Potassium Phosphate, Monobasic, Molecular Biology Grade, Calbiochem™, , 1kg

Catalog No. 529568250GM
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
529568250GM 250 g
5295681KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 529568250GM Supplier MilliporeSigma™ Supplier No. 529568250GM

Specifications

Chemical Name or Material Potassium Phosphate, Monobasic
CAS 7778-77-0
Quantity 250 g
Purity Grade Notes by titration
Assay Percent Range ≥99%
Molecular Formula H2KO4P
MDL Number MFCD00011401 MFCD00147253
Synonym potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
SMILES [K+].OP(O)([O-])=O
IUPAC Name potassium dihydrogen phosphate
Molecular Weight (g/mol) 136.08
PubChem CID 516951
ChEBI CHEBI:63036
Formula Weight 136.1
Grade Molecular Biology
Show More Show Less