missing translation for 'onlineSavingsMsg'
Learn More

Potassium Phosphate Dibasic, (White crystalline powder/ACS Reagent Grade), MP Biomedicals

Catalog No. ICN19143105
Change view
Click to view available options
Quantity:
1 kg
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19143105 5 kg
ICN19143101 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19143105 Supplier MP Biomedicals Inc Supplier No. 0219143105

Chemical Identifiers

CAS 11-4-7758
Molecular Formula HK2O4P
Molecular Weight (g/mol) 174.17
InChI Key ZPWVASYFFYYZEW-UHFFFAOYSA-L
Synonym dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate
PubChem CID 24450
ChEBI CHEBI:32031
IUPAC Name dipotassium;hydrogen phosphate
SMILES OP(=O)([O-])[O-].[K+].[K+]

Specifications

Color White
Physical Form Crystalline Powder
Quantity 5 kg
Formula Weight 174.2
Grade ACS Reagent
Chemical Name or Material Potassium Phosphate Dibasic
Recommended Storage Room Temperature