missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Phosphate, Dibasic, Ultrapure Bioreagent, J.T. Baker™
Description
- For Liquid Chromatography and Molecular Biology applications
Specifications
Specifications
| Physical Form | Powder |
| Quantity | 2.5 kg |
| Formula Weight | 174.18 |
| Percent Purity | 99.4% |
| Grade | Not Available |
| Packaging | Poly bottle |
| Specific Gravity | 1L = 0.00kg |
| Chemical Name or Material | Potassium Phosphate, Dibasic |