missing translation for 'onlineSavingsMsg'
Learn More

Potassium Phosphate, Dibasic, Powder, Ultrapure Bioreagent, J.T. Baker™

Catalog No. 2003823
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
2003823 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 2003823 Supplier Avantor Supplier No. AVR401201

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For Liquid Chromatography and Molecular Biology applications

Chemical Identifiers

CAS 11-4-7758
Molecular Formula HK2O4P
Molecular Weight (g/mol) 174.17
InChI Key ZPWVASYFFYYZEW-UHFFFAOYSA-L
Synonym dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate
PubChem CID 24450
ChEBI CHEBI:32031
IUPAC Name dipotassium;hydrogen phosphate
SMILES OP(=O)([O-])[O-].[K+].[K+]

Specifications

Physical Form Powder
Quantity 500 g
Formula Weight 174.18
Percent Purity 99.8%
Grade Ultrapure Bioreagent
Packaging Poly Bottle
Chemical Name or Material Potassium Phosphate, Dibasic