Learn More
Potassium Phosphate Dibasic (Fine White Crystalline Powder), Fisher BioReagents™
Supplier: Fisher BioReagents FLBP3631
Description
Potassium Phosphate Dibasic (Fine White Crystalline Powder)
This potassium salt is used in buffers for molecular biology and cell culture applications.Specifications
| Potassium Phosphate Dibasic | |
| 11-4-7758 | |
| 8.5 to 9.6 | |
| 1 kg | |
| HK2O4P | |
| dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate | |
| OP(=O)([O-])[O-].[K+].[K+] | |
| 174.17 | |
| CHEBI:32031 | |
| ≥98.0% | |
| 1.0% max. | |
| >180°C |
| 340°C | |
| White | |
| Solid | |
| ≥98.0 % | |
| 15, 7779 | |
| ZPWVASYFFYYZEW-UHFFFAOYSA-L | |
| dipotassium;hydrogen phosphate | |
| 24450 | |
| 174.18 | |
| Pass Test | |
| Poly Bottle |
Chemical Identifiers
| 11-4-7758 | |
| 174.17 | |
| dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate | |
| CHEBI:32031 | |
| OP(=O)([O-])[O-].[K+].[K+] |
| HK2O4P | |
| ZPWVASYFFYYZEW-UHFFFAOYSA-L | |
| 24450 | |
| dipotassium;hydrogen phosphate |
Safety and Handling
CAUTION!
Emergency Overview
May cause skin, eye, and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.
NFPA
Health:1
Flammability:1
Instability:0
Recommended Storage : RT