missing translation for 'onlineSavingsMsg'
Learn More

Potassium Phosphate, Dibasic, Endotoxin Tested, Multi-Compendial, U.S.P., J.T. Baker™

Catalog No. p-5390850
Change view
Click to view available options
Quantity:
500 g
2.5 kg
5 kg
Packaging:
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02003807 500 g Poly Bottle
02003806 2.5 kg Poly Bottle
02003808 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02003807 Supplier Avantor Supplier No. 325001

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 11-4-7758
Molecular Formula HK2O4P
Molecular Weight (g/mol) 174.17
InChI Key ZPWVASYFFYYZEW-UHFFFAOYSA-L
Synonym dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate
PubChem CID 24450
ChEBI CHEBI:32031
IUPAC Name dipotassium;hydrogen phosphate
SMILES OP(=O)([O-])[O-].[K+].[K+]

Specifications

Quantity 500 g
Formula Weight 174.18
Grade Multi-Compendial/USP
Packaging Poly Bottle
Chemical Name or Material Potassium Phosphate, Dibasic, Endotoxin Tested