missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Phosphate, Dibasic, Endotoxin Tested, Multi-Compendial, U.S.P., J.T. Baker™
Specifications
Specifications
| Quantity | 5 kg |
| Formula Weight | 174.18 |
| Grade | Multi-Compendial/USP |
| Packaging | Poly Pail |
| Chemical Name or Material | Potassium Phosphate, Dibasic, Endotoxin Tested |