missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Phosphate, Dibasic, Anhydrous, FCC, 98%, Spectrum™ Chemical
Description
Spectrum™ Chemical Potassium Phosphate, Dibasic, Anhydrous, FCC is used as an acidity regulator, antioxidant, sequestrant and stabilizer in food and beverages. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage, nutritional supplement and personal care applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 1 |
| Physical Form | Crystals |
| Quantity | 500 g |
| Percent Purity | 98% |
| Grade | FCC |
| Loss on Drying | 0.02 |
| Packaging | Poly Bottle |
| Chemical Name or Material | Potassium Phosphate |