Learn More
Potassium Phosphate Dibasic Anhydrous (Crystalline Powder/USP), Fisher Chemical™
Supplier: Fisher Chemical FLP290212
Specifications
| Potassium Phosphate Dibasic Anhydrous | |
| 11-4-7758 | |
| 8.5 to 9.6 | |
| 2.5 kg | |
| MFCD00011383 | |
| ZPWVASYFFYYZEW-UHFFFAOYSA-L | |
| dipotassium;hydrogen phosphate | |
| 24450 | |
| 174.18 | |
| USP | |
| 1.0% max. (USP) | |
| >180°C |
| 340°C | |
| White | |
| Solid | |
| HK2O4P | |
| dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate | |
| OP(=O)([O-])[O-].[K+].[K+] | |
| 174.17 | |
| CHEBI:32031 | |
| 98.0 to 100.5% (USP) | |
| Pass Test (USP) | |
| Poly Bottle |
Chemical Identifiers
| 11-4-7758 | |
| 174.17 | |
| ZPWVASYFFYYZEW-UHFFFAOYSA-L | |
| 24450 | |
| dipotassium;hydrogen phosphate |
| HK2O4P | |
| MFCD00011383 | |
| dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate | |
| CHEBI:32031 | |
| OP(=O)([O-])[O-].[K+].[K+] |
Safety and Handling
CAUTION!
Emergency Overview
May cause skin, eye, and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.
NFPA
Health:1
Flammability:1
Instability:0