missing translation for 'onlineSavingsMsg'
Learn More

Potassium Phosphate Dibasic Anhydrous (Crystalline Powder/USP), Fisher Chemical™

Supplier:  Fisher Chemical FLP29012

 View more versions of this product

Catalog No. P290-12


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Potassium Phosphate Dibasic Anhydrous
11-4-7758
8.5 to 9.6
12 kg
MFCD00011383
ZPWVASYFFYYZEW-UHFFFAOYSA-L
dipotassium;hydrogen phosphate
24450
174.18
USP
1.0% max. (USP)
>180°C
340°C
White
Solid
HK2O4P
dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate
OP(=O)([O-])[O-].[K+].[K+]
174.17
CHEBI:32031
98.0 to 100.5% (USP)
Pass Test (USP)
Poly Pail

Chemical Identifiers

11-4-7758
174.17
ZPWVASYFFYYZEW-UHFFFAOYSA-L
24450
dipotassium;hydrogen phosphate
HK2O4P
MFCD00011383
dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate
CHEBI:32031
OP(=O)([O-])[O-].[K+].[K+]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

CAUTION!

Emergency Overview
May cause skin, eye, and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:1
Flammability:1
Instability:0

SDS Documents

Documents

Promotions

Promotions