missing translation for 'onlineSavingsMsg'
Learn More

Potassium Phosphate, Dibasic, ACS Grade, ≥ 98.0%, LabChem™

Catalog No. LC200901
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC200901 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC200901 Supplier LabChem Supplier No. LC200901

Chemical Identifiers

CAS 11-4-7758
Molecular Formula HK2O4P
Molecular Weight (g/mol) 174.17
InChI Key ZPWVASYFFYYZEW-UHFFFAOYSA-L
Synonym dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate
PubChem CID 24450
ChEBI CHEBI:32031
IUPAC Name dipotassium;hydrogen phosphate
SMILES OP(=O)([O-])[O-].[K+].[K+]

Specifications

Melting Point 340°C
Color White
CAS Max % 100
Physical Form Granules
Quantity 500 g
Linear Formula K2HPO4
Solubility Information Soluble in water
Formula Weight 174.18
Percent Purity ≥98.0%
Grade ACS
Identification Passes Test
Packaging Poly Bottle
Decomposition Information Phosphorus oxides
Chemical Name or Material Potassium Phosphate, Dibasic
Show More Show Less
Hazard Statement GHS H Statement
Product is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
EINECSNumber 231-834-5
Recommended Storage Room Temperature