missing translation for 'onlineSavingsMsg'
Learn More

Potassium phosphate, dibasic, 99%, for HPLC, anhydrous

Catalog No. AC447362500
Change view
Click to view available options
Quantity:
250 g
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC447362500 250 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC447362500 Supplier Thermo Scientific Chemicals Supplier No. 447362500
Get 1 Extra Chemical When You Buy 2
View all promotions

CAS: 7758-11-4 | HK2O4P | 174.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For HPLC

Chemical Identifiers

CAS 11-4-7758
Molecular Formula HK2O4P
Molecular Weight (g/mol) 174.18
InChI Key ZPWVASYFFYYZEW-UHFFFAOYSA-L
Synonym dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate
PubChem CID 24450
ChEBI CHEBI:32031
IUPAC Name dipotassium;hydrogen phosphate
SMILES OP(=O)([O-])[O-].[K+].[K+]

Specifications

Melting Point 340.0°C
Quantity 250 g
Assay Percent Range 99%
Linear Formula K2HPO4
Merck Index 15,7779
Solubility Information Clear colorless
Formula Weight 174.18
Percent Purity 99%
Grade HPLC
Packaging Plastic bottle
Chemical Name or Material Potassium phosphate, dibasic
Show More Show Less
EINECSNumber 231-834-5

RUO – Research Use Only