missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Phosphate Dibasic, ≥98%, ACS reagent, Honeywell Fluka™
ACS Reagent,≥98%
Supplier: Honeywell Chemicals P378625KG
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
| Potassium phosphate dibasic | |
| 2.5 kg | |
| K2HPO4 | |
| NONH for all modes of transport | |
| ZPWVASYFFYYZEW-UHFFFAOYSA-L | |
| dipotassium;hydrogen phosphate | |
| 24450 | |
| 174.18g/mol |
| 11-4-7758 | |
| HK2O4P | |
| MFCD00011383 | |
| dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate | |
| OP(=O)([O-])[O-].[K+].[K+] | |
| 174.17 | |
| CHEBI:32031 | |
| ≥98% |
Chemical Identifiers
| 11-4-7758 | |
| 174.17 | |
| ZPWVASYFFYYZEW-UHFFFAOYSA-L | |
| 24450 | |
| dipotassium;hydrogen phosphate |
| HK2O4P | |
| MFCD00011383 | |
| dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate | |
| CHEBI:32031 | |
| OP(=O)([O-])[O-].[K+].[K+] |