missing translation for 'onlineSavingsMsg'
Learn More

Potassium phosphate, anhydrous, 97%

Catalog No. AA41587A7
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41587A7 5 kg
AA4158730 250 g
AA41587A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA41587A7 Supplier Thermo Scientific Chemicals Supplier No. 041587A7

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7778-53-2 | K3O4P | 212.265 g/mol

Potassium phosphate is used as a fertilizer and food additive. It serves as a buffering agent to regulate pH. It is also used in optical modulators and for non-linear optics such as second-harmonic generation (SHG). It is involved in the phase-transfer catalyzed preparation of acetylenes from the corresponding vicinal dibromo compounds through double dehydrobromination. It is also useful for the oxidative coupling of thiols to disulfide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7778-53-2
Molecular Formula K3O4P
Molecular Weight (g/mol) 212.265
MDL Number MFCD00036295
InChI Key LWIHDJKSTIGBAC-UHFFFAOYSA-K
Synonym potassium phosphate, tripotassium phosphate, potassium phosphate tribasic, phosphoric acid, tripotassium salt, potassium orthophosphate, tripotassium orthophosphate, caswell no. 700a, potassium phosphate, tribasic, potassium phosphate k3po4, ccris 7321
PubChem CID 62657
IUPAC Name tripotassium;phosphate
SMILES [O-]P(=O)([O-])[O-].[K+].[K+].[K+]

Specifications

Melting Point 1340°C
Physical Form Granules
Quantity 5 kg
Merck Index 14,7660
Solubility Information Soluble in water; Insoluble in alcohol.
Formula Weight 212.27
Percent Purity 97%
Sensitivity Hygroscopic
Density 2.564
Chemical Name or Material Potassium phosphate, anhydrous
Hazard Category H318-H335
Hazard Statement GHS H Statement
H315-H319;
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P271-P280-P304+P340-P305+P351+P338-P310-P312-P501c
EINECSNumber 231-907-1
RTECSNumber TC8450000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only