missing translation for 'onlineSavingsMsg'
Learn More

Potassium phosphate, ACS, 98.0% min

Catalog No. p-7080166
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4664430 250 g
AA46644A1 1 kg
AA46644A7 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4664430 Supplier Thermo Scientific Chemicals Supplier No. 04664430
Only null left

CAS: 7778-53-2 | K3O4P | 212.265 g/mol

Potassium phosphate tribasic is a reagent with a very high buffering capacity. It is widely used in molecular biology, biochemistry, and chromatography, used for the extraction of keratohyalin protein from bovine tissue, to study the effects of freezing and thawing on the stability of proteins sensitive to conformational changes. It is used as a food additive for its properties as an emulsifier, foaming agent, and whipping agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7778-53-2
Molecular Formula K3O4P
Molecular Weight (g/mol) 212.265
MDL Number MFCD00036295
InChI Key LWIHDJKSTIGBAC-UHFFFAOYSA-K
Synonym potassium phosphate, tripotassium phosphate, potassium phosphate tribasic, phosphoric acid, tripotassium salt, potassium orthophosphate, tripotassium orthophosphate, caswell no. 700a, potassium phosphate, tribasic, potassium phosphate k3po4, ccris 7321
PubChem CID 62657
IUPAC Name tripotassium;phosphate
SMILES [O-]P(=O)([O-])[O-].[K+].[K+].[K+]

Specifications

Melting Point 1340°C
Color White
Physical Form Granules
Quantity 250 g
Merck Index 14,7660
Solubility Information Soluble in water; Insoluble in alcohol;
Formula Weight 212.27
Percent Purity ≥98.0%
Grade ACS
Sensitivity Hygroscopic
Density 2.564 g/mL
Chemical Name or Material Potassium phosphate
Show More Show Less
Hazard Category H318-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P271-P280-P304+P340-P305+P351+P338-P310-P312-P501c
EINECSNumber 231-907-1
RTECSNumber TC8450000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only