missing translation for 'onlineSavingsMsg'
Learn More

Potassium Phenyl Sulfate 98.0+%, TCI America™

Catalog No. p-7134505
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P02321G 1 g
P02325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P02321G Supplier TCI America Supplier No. P02321G

Chemical Identifiers

CAS 1733-88-6
Molecular Formula C6H5KO4S
Molecular Weight (g/mol) 212.26
InChI Key NOUFXYHWAWIDNT-UHFFFAOYSA-M
Synonym Sulfuric Acid Phenyl Ester Potassium Salt
PubChem CID 23678854
IUPAC Name potassium;phenyl sulfate
SMILES C1=CC=C(C=C1)OS(=O)(=O)[O-].[K+]

Specifications

Melting Point 160°C
Color White
Quantity 1 g
Formula Weight 212.26
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Potassium Phenyl Sulfate
RTECSNumber WT1180000
TSCA No
Recommended Storage Freezer