missing translation for 'onlineSavingsMsg'
Learn More

Potassium Persulfate, Crystal, 99%, Spectrum™ Chemical

Catalog No. 18612874
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612874 50 kg Poly Bottle
18612870 125 g Poly Bottle
18612871 500 g Poly Bottle
18612872 2.5 kg Poly Bottle
18612873 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18612874 Supplier Spectrum Chemical Mfg Cor Supplier No. P137550KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7727-21-1 | Weight: 270.32 g/mol

Chemical Identifiers

CAS 7727-21-1

Specifications

Physical Form Crystalline Powder or Needles
CAS Max % 1
Quantity 50 kg
Packaging Poly Bottle
Molecular Weight (g/mol) 270.32
Percent Purity 99%
Grade Reagent
Chemical Name or Material Potassium Persulfate, Crystal

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.