missing translation for 'onlineSavingsMsg'
Learn More

Potassium Persulfate, ACS Reagent (Potassium Peroxydisulfate), Reagents

Catalog No. 01337807
Change view
Click to view available options
Quantity:
125 g
500 g
1 kg
10 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337807 10 kg
01337804 500 g
01337806 125 g
01337805 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01337807 Supplier Labchem Inc Supplier No. C128860010KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Potassium Persulfate, ACS Reagent (Potassium Peroxydisulfate), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7727-21-1
Molecular Formula K2O8S2
Molecular Weight (g/mol) 270.31
InChI Key USHAGKDGDHPEEY-UHFFFAOYSA-L
Synonym Potassium Peroxydisulfate, Potassium Peroxodisulphate
IUPAC Name dipotassium [(sulfonatoperoxy)sulfonyl]oxidanide
SMILES [K+].[K+].[O-]S(=O)(=O)OOS([O-])(=O)=O

Specifications

Melting Point >170°C
Color Colorless/White
Physical Form Solid
Quantity 10 kg
Percent Purity 99.8%
Grade ACS
Packaging Poly Pail
Specific Gravity 2.477
Chemical Name or Material Potassium Persulfate, ACS Reagent (Potassium Peroxydisulfate)