missing translation for 'onlineSavingsMsg'
Learn More

Potassium persulfate, 99+%, ACS reagent

CAS: 7727-21-1 | K2O8S2 | 270.3 g/mol

Supplier:  Thermo Scientific Chemicals 424180025

 View more versions of this product

Catalog No. AC424180025

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Oxidation reagent
Specifications

Specifications

Potassium persulfate
99+%
White
2.5 kg
K2O8S2
MFCD00011386
15, 7777
Solubility in water: 5g/100mL (20°C). Other solubilities: insoluble in alcohol
[O-]S(=O)(=O)OOS(=O)(=O)[O-].[K+].[K+]
270.3
270.3
ACS Reagent
2.47
100.0°C
7727-21-1
Fine Hygroscopic Crystalline Powder
99+%
K2S2O8
08,417; 15,274
potassium persulfate, anthion, potassium peroxydisulfate, potassium peroxodisulfate, dipotassium peroxydisulfate, potassium peroxydisulphate, dipotassium persulfate, caswell no. 700, dipotassium peroxodisulphate, unii-6b86k0mczc
USHAGKDGDHPEEY-UHFFFAOYSA-L
dipotassium;sulfonatooxy sulfate
24412
≥99%
Plastic bottle

Chemical Identifiers

7727-21-1
270.3
USHAGKDGDHPEEY-UHFFFAOYSA-L
24412
[O-]S(=O)(=O)OOS(=O)(=O)[O-].[K+].[K+]
K2O8S2
MFCD00011386
potassium persulfate, anthion, potassium peroxydisulfate, potassium peroxodisulfate, dipotassium peroxydisulfate, potassium peroxydisulphate, dipotassium persulfate, caswell no. 700, dipotassium peroxodisulphate, unii-6b86k0mczc
dipotassium;sulfonatooxy sulfate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
May intensify fire; oxidizer.
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
May cause allergy or asthma symptoms or breathing difficu

GHS P Statement
Keep/Store away from clothing/ combustible materials.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,r

GHS Signal Word: Danger

EINECSNumber : 231-781-8

RTECSNumber : SE0400000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only