missing translation for 'onlineSavingsMsg'
Learn More

Potassium Perrhenate, TCI America™

Catalog No. p19391g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P19391G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P19391G Supplier TCI America Supplier No. P19391G
Only null left

Chemical Identifiers

CAS 10466-65-6
Molecular Formula KO4Re
Molecular Weight (g/mol) 289.30
MDL Number MFCD00011365
InChI Key QFKRWIFGDGKWLM-UHFFFAOYSA-N
Synonym potassium perrhenate, potassiumperrhenate, k.o4re, potassium rhenium oxide kreo4, perrhenic acid hreo4 , potassium salt, potassium perrhenate vii, acmc-1bx96, perrhenic acid potassium salt, potassium oxido trioxo rhenium, potassium perrhenate kreo4
PubChem CID 3661560
IUPAC Name potassium rheniumoylolate
SMILES [K+].[O-][Re](=O)(=O)=O

Specifications

Melting Point 550°C
Color Gray-White
Physical Form Crystalline Powder
Quantity 1 g
UN Number 1479
Formula Weight 289.30
Chemical Name or Material Potassium Perrhenate
RTECSNumber SE1400000
TSCA Yes