missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium perrhenate, 99% (metals basis), Re 64%
Description
A powerful oxidizer. Also used for spectrometry, tungsten wire cathode coating color film and as catalyst. Potassium perrhenate is used as strong oxidizing agent and used in the synthesis of technetium and rhenium organometallic radiopharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 1360°C to 1370°C |
| Melting Point | 550°C |
| Color | White |
| Physical Form | Crystalline |
| Quantity | 10 g |
| UN Number | UN1479 |
| Solubility Information | Soluble in water. |
| Formula Weight | 289.3 |
| Percent Purity | 99% |
| Concentration or Composition (by Analyte or Components) | Re 64% |
| Show More |
Safety and Handling
| Hazard Category | H272-H315-H319-H335 |
| Hazard Statement | GHS H Statement H272 May intensify fire oxidizer. |
| Precautionary Statement | P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S. |
| EINECSNumber | 233-953-8 |
| RTECSNumber | SE1400000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only