missing translation for 'onlineSavingsMsg'
Learn More

Potassium perrhenate, 99% (metals basis), Re 64%

Catalog No. AA1141104
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1141104 2 g
AA1141109 10 g
AA1141118 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1141104 Supplier Thermo Scientific Chemicals Supplier No. 01141104
Only null left

A powerful oxidizer | CAS: 10466-65-6 | KO4Re | 289.30 g/mol

A powerful oxidizer. Also used for spectrometry, tungsten wire cathode coating color film and as catalyst. Potassium perrhenate is used as strong oxidizing agent and used in the synthesis of technetium and rhenium organometallic radiopharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10466-65-6
Molecular Formula KO4Re
Molecular Weight (g/mol) 289.30
MDL Number MFCD00011365
InChI Key QFKRWIFGDGKWLM-UHFFFAOYSA-N
Synonym potassium perrhenate, potassiumperrhenate, k.o4re, potassium rhenium oxide kreo4, perrhenic acid hreo4 , potassium salt, potassium perrhenate vii, acmc-1bx96, perrhenic acid potassium salt, potassium oxido trioxo rhenium, potassium perrhenate kreo4
PubChem CID 3661560
IUPAC Name potassium;oxido(trioxo)rhenium
SMILES [K+].[O-][Re](=O)(=O)=O

Specifications

Boiling Point 1360°C to 1370°C
Melting Point 550°C
Color White
Physical Form Crystalline
Quantity 2 g
UN Number UN1479
Solubility Information Soluble in water.
Formula Weight 289.3
Percent Purity 99%
Concentration or Composition (by Analyte or Components) Re 64%
Odor Odorless
Refractive Index 1.643
Density 4.887 g/mL
Assay (metals basis)
Chemical Name or Material Potassium perrhenate
Show More Show Less
Hazard Category H272-H315-H319-H335
Hazard Statement GHS H Statement
H272
May intensify fire
oxidizer.
Precautionary Statement P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.
EINECSNumber 233-953-8
RTECSNumber SE1400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only