missing translation for 'onlineSavingsMsg'
Learn More

Potassium Peroxymonosulfate [> ca. 45%(T) as KHSO5], TCI America™

Catalog No. p-7134285
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O031025G 25 g
O0310500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O031025G Supplier TCI America Supplier No. O031025G
Only null left

Chemical Identifiers

CAS 37222-66-5
Molecular Formula H3K5O18S4
Synonym Oxone

Specifications

Color White
pH 2.3 (10g/L H2O soln. 77°C)
Physical Form Crystalline Powder
Quantity 25 g
UN Number 3260
Formula Weight 614.74
Chemical Name or Material Potassium Peroxymonosulfate [> ca. 45%(T) as KHSO5]