Learn More
Potassium peroxydisulfate, 97%
Description
Potassium peroxydisulfate is used as an oxidizing agent in organic synthesis. It is involved in the Elbs persulfate oxidation of phenols and the Boyland-Sims oxidation of anilines. In solution, it gives radicals and is used to initiate polymerization reactions to prepare styrene-butadiene rubber and polytetrafluoroethylene. It is used as a bleaching agent in various hair bleaches, as an ion exchange agent, a plating agent and a surface treating agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Potassium peroxydisulfate is used as an oxidizing agent in organic synthesis. It is involved in the Elbs persulfate oxidation of phenols and the Boyland-Sims oxidation of anilines. In solution, it gives radicals and is used to initiate polymerization reactions to prepare styrene-butadiene rubber and polytetrafluoroethylene. It is used as a bleaching agent in various hair bleaches, as an ion exchange agent, a plating agent and a surface treating agent.
Solubility
Soluble in water. Insoluble in alcohol.
Notes
Moisture sensitive. Incompatible with organic materials, strong reducing agents, powdered metals, strong bases, alcohols, phosphorous, anhydrides, halogens and acids.
Specifications
Specifications
| Melting Point | 100°C (dec.) |
| Quantity | 500 g |
| Odor | Odorless |
| Density | 2.477 |
| Linear Formula | K2S2O8 |
| UN Number | UN1492 |
| Merck Index | 14,7656 |
| Solubility Information | Soluble in water. Insoluble in alcohol. |
| Formula Weight | 270.33 |
| Percent Purity | 97% |
| Show More |
Safety and Handling
| Hazard Category | H272-H302-H315-H317-H319-H334-H335 |
| Hazard Statement | GHS H Statement H334-H272-H302-H335-H315-H319-H317 May cause allergy or asthma symptoms or breathing difficulties if inhaled. May intensify fire; oxidizer. Harmful if swallowed. May cause respiratory irritation. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction. |
| Precautionary Statement | P210-P220-P221-P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P330-P333+P313-P342+P311-P362-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: POTASSIUM PERSULFATE |
| EINECSNumber | 231-781-8 |
| RTECSNumber | SE0400000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only