missing translation for 'onlineSavingsMsg'
Learn More

Potassium Periodate, Technical, 99%, Spectrum™ Chemical

Catalog No. p-7166067
Change view
Click to view available options
Quantity:
2.5 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Catalog No. Quantity Packaging
18612824 2.5 kg Amber Glass Bottle
18612826 50 kg Fiber Drum
2 options

Catalog No. 18612824

Supplier: Spectrum Chemical Mfg Cor P13282.5KG

Only null left

Chemical Identifiers

CAS 7790-21-8

Specifications

CAS Max % 1
Quantity 2.5 kg
Percent Purity 99%
Grade Technical
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.