missing translation for 'onlineSavingsMsg'
Learn More

Potassium Periodate, Technical, 99%, Spectrum™ Chemical

Catalog No. 18612824
Change view
Click to view available options
Quantity:
2.5 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612824 2.5 kg Amber Glass Bottle
18612826 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18612824 Supplier Spectrum Chemical Mfg Cor Supplier No. P13282.5KG
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 7790-21-8

Specifications

CAS Max % 1
Quantity 2.5 kg
Percent Purity 99%
Grade Technical
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.