missing translation for 'onlineSavingsMsg'
Learn More

Potassium Periodate, ACS Reagent (Potassium Metaperiodate), Reagents

Catalog No. 01337813
Change view
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
25 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337813 500 g
01337816 10 kg
01337815 2.5 kg
01337814 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01337813 Supplier Labchem Inc Supplier No. C1288200500A5

Potassium Periodate, ACS Reagent (Potassium Metaperiodate), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7790-21-8
Molecular Formula IKO4
Molecular Weight (g/mol) 230.00
InChI Key FJVZDOGVDJCCCR-UHFFFAOYSA-M
Synonym Potassium Metaperiodate
IUPAC Name potassium periodate
SMILES [K+].[O-][I](=O)(=O)=O

Specifications

Boiling Point 300°C (loses oxygen)
Color White
Physical Form Solid
Quantity 500 g
Grade ACS
Packaging Poly Bottle
Specific Gravity 3.618
Chemical Name or Material Potassium Periodate, ACS Reagent (Potassium Metaperiodate)