Learn More
Potassium perchlorate, anhydrous, ACS, 99.0-100.5%
For Parr Bomb Work | CAS: 7778-74-7 | ClKO4 | 138.54 g/mol
$106.53 - $350.23
Chemical Identifiers
| CAS | 7778-74-7 |
|---|---|
| Molecular Formula | ClKO4 |
| Molecular Weight (g/mol) | 138.54 |
| MDL Number | MFCD00011362 |
| InChI Key | YLMGFJXSLBMXHK-UHFFFAOYSA-M |
| Synonym | potassium perchlorate, perchloracap, astrumal, peroidin, perchloric acid, potassium salt, irenat, potassium hyperchloride, perchloric acid, potassium salt 1:1, potassium perchlorate kclo4, potassium perchlorate usp |
| PubChem CID | 516900 |
| IUPAC Name | potassium;perchlorate |
| SMILES | [K+].[O-][Cl](=O)(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA1163022
|
Thermo Scientific Chemicals
01163022 |
100 g |
Each for $106.53
|
|
|||||
|
AA1163036
|
Thermo Scientific Chemicals
01163036 |
500 g |
N/A
|
N/A | |||||
|
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. |
|||||||||
Description
For Parr Bomb WorkPotassium perchlorate is used in ammunition percussion caps, explosive primers, fireworks and propellants. It acts as a solid rocket propellant. It is also used as an oxidizer in colored pyrotechnic compositions and flash powder. It is used as an antithyroid and involved in the treatment of hyperthyroidism. The mixture of anthracene and sulfur with potassium perchlorate is used for generating black smoke signaling.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7778-74-7 | |
| 138.54 | |
| YLMGFJXSLBMXHK-UHFFFAOYSA-M | |
| 516900 | |
| [K+].[O-][Cl](=O)(=O)=O |
| ClKO4 | |
| MFCD00011362 | |
| potassium perchlorate, perchloracap, astrumal, peroidin, perchloric acid, potassium salt, irenat, potassium hyperchloride, perchloric acid, potassium salt 1:1, potassium perchlorate kclo4, potassium perchlorate usp | |
| potassium;perchlorate |
Specifications
| 400°C (decomposition) | |
| Black | |
| 100 g | |
| ClKO4 | |
| potassium perchlorate, perchloracap, astrumal, peroidin, perchloric acid, potassium salt, irenat, potassium hyperchloride, perchloric acid, potassium salt 1:1, potassium perchlorate kclo4, potassium perchlorate usp | |
| YLMGFJXSLBMXHK-UHFFFAOYSA-M | |
| potassium;perchlorate | |
| 516900 | |
| ACS | |
| Hygroscopic | |
| 2.52 g/cm3 at 20°C |
| 7778-74-7 | |
| Powder | |
| 99.0-100.5% | |
| MFCD00011362 | |
| Partially soluble in water. Insoluble in alcohol.Soluble in water. Slightly soluble in acetone. Insoluble in ethanol,ethyl acetate and ether. | |
| [K+].[O-][Cl](=O)(=O)=O | |
| 138.54 | |
| 138.55 | |
| Odorless | |
| White powder | |
| Potassium perchlorate, anhydrous, For analysis ACS |
Safety and Handling
P210-P220-P221-P264b-P270-P280-P283-P301+P312-P306+P360-P330-P370+P378q-P371+P380+P375-P501c
H271-H302
DOTInformation : DOT Class: 5.1, Packaging Group: II
EINECSNumber : 231-912-9
RTECSNumber : SC9700000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only