missing translation for 'onlineSavingsMsg'
Learn More

Potassium perchlorate, ACS reagent

CAS: 7778-74-7 | ClKO4 | 138.54 g/mol

$98.29 - $98.29

Chemical Identifiers

CAS 7778-74-7
Molecular Formula ClKO4
Molecular Weight (g/mol) 138.54
MDL Number MFCD00011362
InChI Key YLMGFJXSLBMXHK-UHFFFAOYSA-M
Synonym potassium perchlorate, perchloracap, astrumal, peroidin, perchloric acid, potassium salt, irenat, potassium hyperchloride, perchloric acid, potassium salt 1:1, potassium perchlorate kclo4, potassium perchlorate usp
PubChem CID 516900
IUPAC Name potassium;perchlorate
SMILES [K+].[O-][Cl](=O)(=O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC221371000
SDS View Documents
Thermo Scientific Chemicals
221371000
100 g Plastic bottle N/A
N/A
Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7778-74-7
138.54
YLMGFJXSLBMXHK-UHFFFAOYSA-M
516900
[K+].[O-][Cl](=O)(=O)=O
ClKO4
MFCD00011362
potassium perchlorate, perchloracap, astrumal, peroidin, perchloric acid, potassium salt, irenat, potassium hyperchloride, perchloric acid, potassium salt 1:1, potassium perchlorate kclo4, potassium perchlorate usp
potassium;perchlorate

Specifications

400.0°C
100 g
KClO4
15,7774
Solubility in water: 17g/L (20°C). Other solubilities: 0.75 g/100mL water (0°C), 21.8 g/100mL water (100°C), practically insoluble in alcohol
[K+].[O-][Cl](=O)(=O)=O
138.54
138.55
Plastic bottle
7778-74-7
ClKO4
MFCD00011362
potassium perchlorate, perchloracap, astrumal, peroidin, perchloric acid, potassium salt, irenat, potassium hyperchloride, perchloric acid, potassium salt 1:1, potassium perchlorate kclo4, potassium perchlorate usp
YLMGFJXSLBMXHK-UHFFFAOYSA-M
potassium;perchlorate
516900
ACS Reagent
Potassium perchlorate
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

EINECSNumber : 231-912-9

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only