missing translation for 'onlineSavingsMsg'
Learn More

Potassium perchlorate, ACS reagent

Catalog No. AC221371000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC221371000 100 g Plastic bottle
AC221375000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC221371000 Supplier Thermo Scientific Chemicals Supplier No. 221371000

Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7778-74-7 | ClKO4 | 138.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7778-74-7
Molecular Formula ClKO4
Molecular Weight (g/mol) 138.54
MDL Number MFCD00011362
InChI Key YLMGFJXSLBMXHK-UHFFFAOYSA-M
Synonym potassium perchlorate, perchloracap, astrumal, peroidin, perchloric acid, potassium salt, irenat, potassium hyperchloride, perchloric acid, potassium salt 1:1, potassium perchlorate kclo4, potassium perchlorate usp
PubChem CID 516900
IUPAC Name potassium;perchlorate
SMILES [K+].[O-][Cl](=O)(=O)=O

Specifications

Melting Point 400.0°C
Quantity 100 g
Linear Formula KClO4
Merck Index 15,7774
Solubility Information Solubility in water: 17g/L (20°C). Other solubilities: 0.75 g/100mL water (0°C), 21.8 g/100mL water (100°C), practically insoluble in alcohol
Formula Weight 138.55
Grade ACS Reagent
Packaging Plastic bottle
Chemical Name or Material Potassium perchlorate
Hazard Category GHS Signal Word: Danger
EINECSNumber 231-912-9

RUO – Research Use Only