missing translation for 'onlineSavingsMsg'
Learn More

Potassium p-Tolyltrifluoroborate 98.0+%, TCI America™

Catalog No. P15835G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P15835G 5 g
P15831G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P15835G Supplier TCI America Supplier No. P15835G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 216434-82-1
Molecular Formula C7H7BF3K
Molecular Weight (g/mol) 198.037
MDL Number MFCD01318171
InChI Key KRWDYXJWQBTBAH-UHFFFAOYSA-N
Synonym potassium p-tolyltrifluoroborate, potassium trifluoro p-tolyl borate, potassium 4-methylphenyl trifluoroborate, potassium 4-methylphenyltrifluoroborate, potassium trifluoro 4-methylphenyl boranuide, 4-methylphenyltrifluoroboric acid potassium salt, pubchem11452, trifluoro 4-methylphenyl potassioboron v, potassium trifluoro p-tolyl boranuide, potassium trifluoro-4-methylphenyl boranuide
PubChem CID 23682258
IUPAC Name potassium;trifluoro-(4-methylphenyl)boranuide
SMILES [B-](C1=CC=C(C=C1)C)(F)(F)F.[K+]

Specifications

Melting Point 294°C
Color White
Quantity 5 g
Formula Weight 198.04
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Potassium p-Tolyltrifluoroborate