missing translation for 'onlineSavingsMsg'
Learn More

Potassium p-Tolyl Sulfate 98.0+%, TCI America™

Catalog No. P20911G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P20911G 1 g
P20915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P20911G Supplier TCI America Supplier No. P20911G

Chemical Identifiers

CAS 91978-69-7
Molecular Formula C7H7KO4S
Molecular Weight (g/mol) 226.29
MDL Number MFCD28100835
InChI Key HTSFIPMTBJHYFQ-UHFFFAOYSA-M
Synonym Potassium p-Cresyl Sulfate, Sulfuric Acid p-Tolyl Ester Potassium Salt, Sulfuric Acid p-Cresyl Ester Potassium Salt
PubChem CID 91972090
IUPAC Name potassium 4-methylphenyl sulfate
SMILES [K+].CC1=CC=C(OS([O-])(=O)=O)C=C1

Specifications

Color White
Quantity 1 g
Formula Weight 226.29
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Potassium p-Tolyl Sulfate
TSCA No
Recommended Storage Freezer