missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium o-Tolyltrifluoroborate 98.0+%, TCI America™
$212.51 - $713.94
Chemical Identifiers
| CAS | 274257-34-0 |
|---|---|
| Molecular Formula | C7H7BF3K |
| Molecular Weight (g/mol) | 198.04 |
| MDL Number | MFCD03790006 |
| InChI Key | ROMUTKWSUQKVLS-UHFFFAOYSA-N |
| Synonym | potassium trifluoro o-tolyl borate, potassium 2-methylphenyl trifluoroborate, potassium o-tolyltrifluoroborate, potassium trifluoro 2-methylphenyl boranuide, potassium 2-methylphenyltrifluoroborate, pubchem11432, potassiumo-tolyltrifluoroborate, trifluoro 2-methylphenyl potassioboron v, potassium trifluoro-2-methylphenyl boranuide, potassium trifluoro 2-methylphenyl borate 1- |
| PubChem CID | 23674703 |
| IUPAC Name | potassium trifluoro(2-methylphenyl)boranuide |
| SMILES | [K+].CC1=CC=CC=C1[B-](F)(F)F |
Chemical Identifiers
| 274257-34-0 | |
| 198.04 | |
| ROMUTKWSUQKVLS-UHFFFAOYSA-N | |
| 23674703 | |
| [K+].CC1=CC=CC=C1[B-](F)(F)F |
| C7H7BF3K | |
| MFCD03790006 | |
| potassium trifluoro o-tolyl borate, potassium 2-methylphenyl trifluoroborate, potassium o-tolyltrifluoroborate, potassium trifluoro 2-methylphenyl boranuide, potassium 2-methylphenyltrifluoroborate, pubchem11432, potassiumo-tolyltrifluoroborate, trifluoro 2-methylphenyl potassioboron v, potassium trifluoro-2-methylphenyl boranuide, potassium trifluoro 2-methylphenyl borate 1- | |
| potassium trifluoro(2-methylphenyl)boranuide |
Specifications
| 274257-34-0 | |
| White-Yellow | |
| MFCD03790006 | |
| potassium trifluoro o-tolyl borate, potassium 2-methylphenyl trifluoroborate, potassium o-tolyltrifluoroborate, potassium trifluoro 2-methylphenyl boranuide, potassium 2-methylphenyltrifluoroborate, pubchem11432, potassiumo-tolyltrifluoroborate, trifluoro 2-methylphenyl potassioboron v, potassium trifluoro-2-methylphenyl boranuide, potassium trifluoro 2-methylphenyl borate 1- | |
| [K+].CC1=CC=CC=C1[B-](F)(F)F | |
| 198.04 | |
| 198.04 | |
| Crystalline Powder |
| 226°C | |
| C7H7BF3K | |
| 1 g | |
| ROMUTKWSUQKVLS-UHFFFAOYSA-N | |
| potassium trifluoro(2-methylphenyl)boranuide | |
| 23674703 | |
| ≥98.0% (HPLC) | |
| Potassium o-Tolyltrifluoroborate |
Safety and Handling
TSCA : No