missing translation for 'onlineSavingsMsg'
Learn More

Potassium o-Tolyltrifluoroborate 98.0+%, TCI America™

Catalog No. P18185G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P18185G 5 g
P18181G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P18185G Supplier TCI America Supplier No. P18185G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 274257-34-0
Molecular Formula C7H7BF3K
Molecular Weight (g/mol) 198.04
MDL Number MFCD03790006
InChI Key ROMUTKWSUQKVLS-UHFFFAOYSA-N
Synonym potassium trifluoro o-tolyl borate, potassium 2-methylphenyl trifluoroborate, potassium o-tolyltrifluoroborate, potassium trifluoro 2-methylphenyl boranuide, potassium 2-methylphenyltrifluoroborate, pubchem11432, potassiumo-tolyltrifluoroborate, trifluoro 2-methylphenyl potassioboron v, potassium trifluoro-2-methylphenyl boranuide, potassium trifluoro 2-methylphenyl borate 1-
PubChem CID 23674703
IUPAC Name potassium trifluoro(2-methylphenyl)boranuide
SMILES [K+].CC1=CC=CC=C1[B-](F)(F)F

Specifications

Melting Point 226°C
Color White-Yellow
Quantity 5 g
Formula Weight 198.04
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Potassium o-Tolyltrifluoroborate