missing translation for 'onlineSavingsMsg'
Learn More

Potassium nitrate, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99%, Honeywell Fluka™

Catalog No. 60045934
Change view
Click to view available options
Quantity:
100 g
1 kg
6 x 1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60045934 6 x 1 kg
6001279 100 g
6001280 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60045934 Supplier Honeywell Inc Supplier No. 312636X1KG

Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7757-79-1
Molecular Formula KNO3
Molecular Weight (g/mol) 101.10
MDL Number MFCD00011409
InChI Key FGIUAXJPYTZDNR-UHFFFAOYSA-N
Synonym potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras
PubChem CID 24434
ChEBI CHEBI:63043
SMILES [K+].[O-][N+]([O-])=O

Specifications

Melting Point 334°C (literature)
CAS Max % 100.0000%
Quantity 6 x 1 kg
Linear Formula KNO3
UN Number UN1486
IUPAC Name potassium nitrate
Formula Weight 101
Percent Purity >99%
Grade ACS Reagent/Reagent ISO/Reagent Ph. Eur.
Packaging Poly Bottle
Density 2.11 g/cm3
Chemical Name or Material Potassium nitrate, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99%
Show More Show Less
ShelfLife 1260 days from date of manufacture
DOTInformation Transport Hazard Class: 5.1; Packing Group:3: III; Proper Shipping Name: Potassium nitrate
EINECSNumber 231-818-8
Recommended Storage Room Temp